2-Bromo-4-iodo-5-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

164514-1G 164514-500MG 164514-5G 164514-25G
77465-310EA 122.57 USD
77465-310 77478-072 77488-236 77498-920
2-Bromo-4-iodo-5-methylpyridine ≥95%
2-Bromo-4-iodo-5-methylpyridine

Precaución: For research use only

Formula: C₆H₅BrIN
Storage Temperature: Ambient
MDL Number: MFCD16610046
CAS Number: 1227577-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR