3-Bromo-2-(methoxymethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16441-1G 16441-500MG 16441-5G 16441-25G
77465-306EA 370.8 USD
77465-306 77478-068 77488-232 77498-916
3-Bromo-2-(methoxymethoxy)benzaldehyde ≥97%
3-Bromo-2-(methoxymethoxy)benzaldehyde

Precaución: For research use only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21195363
CAS Number: 236094-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR