2-Chloro-4-fluoro-1-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

16389-1G 16389-500MG 16389-5G 16389-25G
77465-302EA 255.44 USD
77465-302 77478-064 77488-228 77498-910
2-Chloro-4-fluoro-1-(methoxymethoxy)benzene ≥95%
2-Chloro-4-fluoro-1-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₈ClFO₂
MW: 190.599 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31376768
CAS Number: 1864105-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR