2,2,2-trifluoro-1-(2-fluoro-5-methoxy-3-(methylthio)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

163704-1G 163704-500MG 163704-5G 163704-25G
77465-298EA 415.09 USD
77465-298 77478-060 77488-226 77498-906
2,2,2-trifluoro-1-(2-fluoro-5-methoxy-3-(methylthio)phenyl)ethanone ≥95%
2,2,2-trifluoro-1-(2-fluoro-5-methoxy-3-(methylthio)phenyl)ethanone

Precaución: For research use only

Formula: C₁₀H₈F₄O₂S
MW: 268.228 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34850196
CAS Number: 2921910-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR