5-Methyl-2-(phenylmethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

16362-1G 16362-500MG 16362-5G 16362-25G
77465-294EA 138.02 USD
77465-294 77478-056 77488-222 77498-904
5-Methyl-2-(phenylmethoxy)benzaldehyde ≥95%
5-Methyl-2-(phenylmethoxy)benzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₄O₂
MW: 226.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06625273
CAS Number: 53389-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR