(2-Bromo-3-chloro-6-fluorophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

163452-1G 163452-500MG 163452-5G 163452-25G
77465-290EA 415.09 USD
77465-290 77478-052 77488-218 77498-898
(2-Bromo-3-chloro-6-fluorophenyl)(methyl)sulfane ≥95%
(2-Bromo-3-chloro-6-fluorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₇H₅BrClFS
Storage Temperature: Ambient
MDL Number: MFCD28141962
CAS Number: 1823313-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR