6-Bromo-2,4-difluoro-3-(trimethylsilyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16290-1G 16290-500MG 16290-5G
77465-286EA 740.57 USD
77465-286 77478-048 77488-214
6-Bromo-2,4-difluoro-3-(trimethylsilyl)benzaldehyde ≥97%
6-Bromo-2,4-difluoro-3-(trimethylsilyl)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁BrF₂OSi
MW: 293.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18642936
CAS Number: 651326-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR