1,5-Dichloro-2-iodo-3-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

162852-1G 162852-500MG 162852-5G
77465-282EA 716.88 USD
77465-282 77478-044 77488-210
1,5-Dichloro-2-iodo-3-(trifluoromethyl)benzene ≥95%
1,5-Dichloro-2-iodo-3-(trifluoromethyl)benzene

Precaución: For research use only

Formula: C₇H₂Cl₂F₃I
MW: 340.896 g/mol
Storage Temperature: Ambient
CAS Number: 2206265-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR