4-Bromo-3-chloro-2-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

162776-1G 162776-500MG 162776-5G
77465-278EA 560.32 USD
77465-278 77478-040 77488-206
4-Bromo-3-chloro-2-hydroxybenzaldehyde ≥97%
4-Bromo-3-chloro-2-hydroxybenzaldehyde

Precaución: For research use only

Formula: C₇H₄BrClO₂
MW: 235.462 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23705565
CAS Number: 1427438-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR