3,5-Dibromo-1H-pyrazole-4-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

162245-1G 162245-500MG 162245-5G
77465-270EA 556.2 USD
77465-270 77478-034 77488-198
3,5-Dibromo-1H-pyrazole-4-carboxylic acid ≥95%
3,5-Dibromo-1H-pyrazole-4-carboxylic acid

Precaución: For research use only

Formula: C₄H₂Br₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00462185
CAS Number: 98026-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR