4-Bromo-3-chloro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

162004-1G 162004-500MG 162004-5G 162004-25G
77465-266EA 336.81 USD
77465-266 77478-030 77488-194 77498-886
4-Bromo-3-chloro-2-methoxybenzaldehyde ≥97%
4-Bromo-3-chloro-2-methoxybenzaldehyde

Precaución: For research use only

Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34849045
CAS Number: 1782181-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR