4-Bromo-3-chloro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

162001-1G 162001-500MG 162001-5G
77465-262EA 518.09 USD
77465-262 77478-026 77488-190
4-Bromo-3-chloro-2-methoxybenzoic acid ≥97%
4-Bromo-3-chloro-2-methoxybenzoic acid

Precaución: For research use only

Formula: C₈H₆BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD34849043
CAS Number: 1783353-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR