3-Bromo-2-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

16176-1G 16176-5G 16176-25G
77465-258EA 64.89 USD
77465-258 77488-184 77498-878
3-Bromo-2-fluoro-4-iodopyridine ≥97%
3-Bromo-2-fluoro-4-iodopyridine

Precaución: For research use only

Formula: C₅H₂BrFIN
Storage Temperature: Ambient
MDL Number: MFCD03095311
CAS Number: 884494-52-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR