Ethyl(4-methoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

161566-1G 161566-500MG 161566-5G 161566-25G
77465-254EA 111.24 USD
77465-254 77478-020 77488-180 77498-874
Ethyl(4-methoxyphenyl)sulfane ≥95%
Ethyl(4-methoxyphenyl)sulfane

Precaución: For research use only

Formula: C₉H₁₂OS
Storage Temperature: Ambient
MDL Number: MFCD11617692
CAS Number: 7205-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR