Ethyl(2-fluoro-5-methoxyphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

161563-1G 161563-500MG 161563-5G 161563-25G
77465-250EA 415.09 USD
77465-250 77478-016 77488-176 77498-870
Ethyl(2-fluoro-5-methoxyphenyl)sulfane ≥95%
Ethyl(2-fluoro-5-methoxyphenyl)sulfane

Precaución: For research use only

Formula: C₉H₁₁FOS
Storage Temperature: Ambient
MDL Number: MFCD22483959
CAS Number: 1824659-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR