(2-Bromo-6-fluorophenyl)(ethyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

161560-1G 161560-500MG 161560-5G
77465-246EA 576.8 USD
77465-246 77478-012 77488-172
(2-Bromo-6-fluorophenyl)(ethyl)sulfane ≥97%
(2-Bromo-6-fluorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₈BrFS
Storage Temperature: Ambient
MDL Number: MFCD22488452
CAS Number: 1822662-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR