2-(Benzo[d][1,3]dioxol-5-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

161552-1G 161552-500MG 161552-5G 161552-25G
77465-242EA 352.26 USD
77465-242 77478-008 77488-166 77498-862
2-(Benzo[d][1,3]dioxol-5-yl)propan-2-ol ≥95%
2-(Benzo[d][1,3]dioxol-5-yl)propan-2-ol

Precaución: For research use only

Formula: C₁₀H₁₂O₃
MW: 180.2004 g/mol
Storage Temperature: Ambient
CAS Number: 119055-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR