(2-Chlorophenyl)(ethyl)sulfane ≥96%

Supplier: AOBChem USA

Warning

161541-1G 161541-500MG 161541-5G 161541-25G
77465-238EA 88.58 USD
77465-238 77478-004 77488-162 77498-858
(2-Chlorophenyl)(ethyl)sulfane ≥96%
(2-Chlorophenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₈H₉ClS
Storage Temperature: Ambient
MDL Number: MFCD00040125
CAS Number: 34126-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR