2-(1-(2,6-Difluorophenyl)vinyl)thiophene ≥95%

Supplier: AOBChem USA

Warning

161017-1G 161017-500MG 161017-5G 161017-25G
77465-234EA 415.09 USD
77465-234 77478-000 77488-156 77498-850
2-(1-(2,6-Difluorophenyl)vinyl)thiophene ≥95%
2-(1-(2,6-Difluorophenyl)vinyl)thiophene

Precaución: For research use only

Formula: C₁₂H₈F₂S
Storage Temperature: Ambient
MDL Number: MFCD34848349
CAS Number: 2921709-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR