3-Bromo-2,6-dimethoxytoluene ≥95%

Supplier: AOBChem USA

Warning

16085-1G 16085-500MG 16085-5G 16085-25G
77465-230EA 92.7 USD
77465-230 77477-996 77488-150 77498-844
3-Bromo-2,6-dimethoxytoluene ≥95%
3-Bromo-2,6-dimethoxytoluene

Precaución: For research use only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD11046750
CAS Number: 22794-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR