5-Bromo-2-(cyclopropylmethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

160682-1G 160682-500MG 160682-5G 160682-25G
77465-226EA 369.77 USD
77465-226 77477-992 77488-146 77498-840
5-Bromo-2-(cyclopropylmethoxy)benzaldehyde ≥95%
5-Bromo-2-(cyclopropylmethoxy)benzaldehyde

Precaución: For research use only

Formula: C₁₁H₁₁BrO₂
MW: 255.108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12138838
CAS Number: 1156170-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR