1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160399-1G 160399-500MG 160399-5G 160399-25G
77465-210EA 415.09 USD
77465-210 77477-976 77488-130 77498-826
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(3-(Benzyloxy)-2-bromo-6-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₈H₂₀BrFO₂
MW: 367.253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34848007
CAS Number: 2921894-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR