1-(2-Bromo-3-chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160396-1G 160396-500MG 160396-5G 160396-25G
77465-206EA 415.09 USD
77465-206 77477-972 77488-126 77498-822
1-(2-Bromo-3-chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Bromo-3-chloro-6-fluorophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃BrClFO
MW: 295.576 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34848004
CAS Number: 2921891-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR