1-(2,6-Difluoro-3-iodophenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160384-1G 160384-500MG 160384-5G 160384-25G
77465-202EA 415.09 USD
77465-202 77477-968 77488-122 77498-818
1-(2,6-Difluoro-3-iodophenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2,6-Difluoro-3-iodophenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₁H₁₃F₂IO
MW: 326.122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34847993
CAS Number: 2921848-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR