1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160364-1G 160364-500MG 160364-5G 160364-25G
77465-198EA 415.09 USD
77465-198 77477-964 77488-118 77498-814
1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(2-Chloro-6-fluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆ClFO
MW: 230.706 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34847973
CAS Number: 2921756-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR