1-(2-Fluoro-5-methylphenyl)-2,2-dimethylpropan-1-ol ≥97%

Supplier: AOBChem USA

Warning

160344-1G 160344-500MG 160344-5G 160344-25G
77465-190EA 415.09 USD
77465-190 77477-956 77488-110 77498-806
1-(2-Fluoro-5-methylphenyl)-2,2-dimethylpropan-1-ol ≥97%
1-(2-Fluoro-5-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇FO
MW: 196.261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17222677
CAS Number: 1342662-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR