1-(5-Bromo-2-fluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

160339-1G 160339-500MG 160339-5G 160339-25G
77465-186EA 415.09 USD
77465-186 77477-952 77488-106 77498-802
1-(5-Bromo-2-fluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(5-Bromo-2-fluoro-3-methylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₆BrFO
MW: 275.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34847950
CAS Number: 2921745-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR