Pyridin-3-yl diethylcarbamate ≥97%

Supplier: AOBChem USA

Warning

16020-1G 16020-500MG 16020-5G 16020-25G
77465-182EA 91.67 USD
77465-182 77477-948 77488-102 77498-800
Pyridin-3-yl diethylcarbamate ≥97%
Pyridin-3-yl diethylcarbamate

Precaución: For research use only

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788469
CAS Number: 51581-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR