2-(2,6-dibromo-4-fluorophenyl)-1,3-dioxolane ≥97%

Supplier: AOBChem USA

Warning

15912-1G 15912-500MG 15912-5G 15912-25G
77465-174EA 278.1 USD
77465-174 77477-940 77488-094 77498-792
2-(2,6-dibromo-4-fluorophenyl)-1,3-dioxolane ≥97%
2-(2,6-dibromo-4-fluorophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₇Br₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD28976845
CAS Number: 2694026-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR