2-Methoxy-6-trifluoromethyl-pyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

15815-1G 15815-500MG 15815-5G 15815-25G
77465-170EA 322.39 USD
77465-170 77477-936 77488-090 77498-790
2-Methoxy-6-trifluoromethyl-pyridine-3-carbaldehyde ≥97%
2-Methoxy-6-trifluoromethyl-pyridine-3-carbaldehyde

Precaución: For research use only

Formula: C₈H₆F₃NO₂
MW: 205.134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696214
CAS Number: 944904-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR