1,3-Dichloro-4-ethoxy-2-iodobenzene ≥95%

Supplier: AOBChem USA

Warning

157647-1G 157647-500MG 157647-5G 157647-25G
77465-166EA 415.09 USD
77465-166 77477-932 77488-080 77498-780
1,3-Dichloro-4-ethoxy-2-iodobenzene ≥95%
1,3-Dichloro-4-ethoxy-2-iodobenzene

Precaución: For research use only

Formula: C₈H₇Cl₂IO
Storage Temperature: Ambient
MDL Number: MFCD34846313
CAS Number: 2384384-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR