2,6-Dichloro-3-ethoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

157639-1G 157639-500MG 157639-5G 157639-25G
77465-162EA 397.58 USD
77465-162 77477-928 77488-076 77498-776
2,6-Dichloro-3-ethoxybenzoic acid ≥95%
2,6-Dichloro-3-ethoxybenzoic acid

Precaución: For research use only

Formula: C₉H₈Cl₂O₃
MW: 235.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28360090
CAS Number: 1936115-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR