1-(Phenylethynyl)cyclohexanol ≥95%

Supplier: AOBChem USA

Warning

157631-1G 157631-5G 157631-25G
77465-158EA 63.86 USD
77465-158 77488-072 77498-772
1-(Phenylethynyl)cyclohexanol ≥95%
1-(Phenylethynyl)cyclohexanol

Precaución: For research use only

Formula: C₁₄H₁₆O
MW: 200.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060883
CAS Number: 20109-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR