(3-(Benzyloxy)-2,6-dichlorophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

157596-1G 157596-500MG 157596-5G 157596-25G
77465-154EA 415.09 USD
77465-154 77477-922 77488-068 77498-768
(3-(Benzyloxy)-2,6-dichlorophenyl)(methyl)sulfane ≥95%
(3-(Benzyloxy)-2,6-dichlorophenyl)(methyl)sulfane

Precaución: For research use only

Formula: C₁₄H₁₂Cl₂OS
Storage Temperature: Ambient
MDL Number: MFCD28360150
CAS Number: 1936630-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR