2-(5-Bromo-2-fluorophenyl)-2-methyl-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

15690-1G 15690-500MG 15690-5G 15690-25G
77465-150EA 94.76 USD
77465-150 77477-918 77488-064 77498-762
2-(5-Bromo-2-fluorophenyl)-2-methyl-1,3-dioxolane ≥95%
2-(5-Bromo-2-fluorophenyl)-2-methyl-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28976831
CAS Number: 1555385-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR