4-(2-Fluoropyridin-3-yl)-1-methylpiperidin-4-ol ≥96%

Supplier: AOBChem USA

Warning

156499-1G 156499-500MG 156499-5G 156499-25G
77465-146EA 415.09 USD
77465-146 77477-914 77488-060 77498-760
4-(2-Fluoropyridin-3-yl)-1-methylpiperidin-4-ol ≥96%
4-(2-Fluoropyridin-3-yl)-1-methylpiperidin-4-ol

Precaución: For research use only

Formula: C₁₁H₁₅FN₂O
Storage Temperature: Ambient
MDL Number: MFCD34845450
CAS Number: 2969429-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR