4-(2-Fluoropyridin-3-yl)tetrahydro-2H-pyran-4-ol ≥97%

Supplier: AOBChem USA

Warning

156495-1G 156495-500MG 156495-5G
77465-142EA 590.19 USD
77465-142 77477-910 77488-056
4-(2-Fluoropyridin-3-yl)tetrahydro-2H-pyran-4-ol ≥97%
4-(2-Fluoropyridin-3-yl)tetrahydro-2H-pyran-4-ol

Precaución: For research use only

Formula: C₁₀H₁₂FNO₂
Storage Temperature: Ambient
MDL Number: MFCD28402119
CAS Number: 1227177-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR