tert-Butyl 4-hydroxy-4-(3-isopropylphenyl)piperidine-1-carboxylate ≥95%

Supplier: AOBChem USA

Warning

155798-1G 155798-500MG 155798-5G 155798-25G
77465-126EA 415.09 USD
77465-126 77477-894 77488-040 77498-742
tert-Butyl 4-hydroxy-4-(3-isopropylphenyl)piperidine-1-carboxylate ≥95%
tert-Butyl 4-hydroxy-4-(3-isopropylphenyl)piperidine-1-carboxylate

Precaución: For research use only

Formula: C₁₉H₂₉NO₃
MW: 319.438 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34845001
CAS Number: 2921882-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR