1-Bromo-3-fluoro-5-(iodomethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

155709-1G 155709-500MG 155709-5G 155709-25G
77465-122EA 397.58 USD
77465-122 77477-890 77488-034 77498-736
1-Bromo-3-fluoro-5-(iodomethyl)benzene ≥97%
1-Bromo-3-fluoro-5-(iodomethyl)benzene

Precaución: For research use only

Formula: C₇H₅BrFI
Storage Temperature: Ambient
MDL Number: MFCD23751968
CAS Number: 1511573-48-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR