Methyl(perfluorophenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

155594-1G 155594-500MG 155594-5G
77465-118EA 715.85 USD
77465-118 77477-886 77488-030
Methyl(perfluorophenyl)sulfane ≥95%
Methyl(perfluorophenyl)sulfane

Precaución: For research use only

Formula: C₇H₃F₅S
Storage Temperature: Ambient
MDL Number: MFCD22053405
CAS Number: 653-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR