2-(Benzo[d][1,3]dioxol-5-yl)cyclopentanol ≥95%

Supplier: AOBChem USA

Warning

154885-1G 154885-500MG 154885-5G 154885-25G
77465-114EA 415.09 USD
77465-114 77477-882 77488-024 77498-718
2-(Benzo[d][1,3]dioxol-5-yl)cyclopentanol ≥95%
2-(Benzo[d][1,3]dioxol-5-yl)cyclopentanol

Precaución: For research use only

Formula: C₁₂H₁₄O₃
Storage Temperature: Ambient
CAS Number: 1824608-03-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR