2,4-Difluoro-1-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

15479-1G 15479-500MG 15479-5G 15479-25G
77465-110EA 197.76 USD
77465-110 77477-878 77488-018 77498-708
2,4-Difluoro-1-(methoxymethoxy)benzene ≥95%
2,4-Difluoro-1-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD25960907
CAS Number: 210350-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR