1,3-Difluoro-5-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

15461-1G 15461-500MG 15461-5G 15461-25G
77465-106EA 219.39 USD
77465-106 77477-874 77488-012 77498-702
1,3-Difluoro-5-(methoxymethoxy)benzene ≥95%
1,3-Difluoro-5-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₈H₈F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD13248876
CAS Number: 749230-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR