1-(3-Bromophenyl)hexan-1-ol ≥97%

Supplier: AOBChem USA

Warning

154586-1G 154586-500MG 154586-5G 154586-25G
77465-102EA 226.6 USD
77465-102 77477-870 77488-008 77498-698
1-(3-Bromophenyl)hexan-1-ol ≥97%
1-(3-Bromophenyl)hexan-1-ol

Precaución: For research use only

Formula: C₁₂H₁₇BrO
MW: 257.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16820775
CAS Number: 71434-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR