1-(3-Bromophenyl)butan-1-ol ≥97%

Supplier: AOBChem USA

Warning

154555-1G 154555-500MG 154555-5G
77465-098EA 471.74 USD
77465-098 77477-866 77488-004
1-(3-Bromophenyl)butan-1-ol ≥97%
1-(3-Bromophenyl)butan-1-ol

Precaución: For research use only

Formula: C₁₀H₁₃BrO
MW: 229.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12153604
CAS Number: 126342-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR