(4-(difluoromethoxy)-3-(trifluoromethyl)phenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

15404-1G 15404-500MG 15404-5G
77465-094EA 638.6 USD
77465-094 77477-862 77488-000
(4-(difluoromethoxy)-3-(trifluoromethyl)phenyl)boronic acid ≥97%
(4-(difluoromethoxy)-3-(trifluoromethyl)phenyl)boronic acid

Precaución: For research use only

Formula: C₈H₆BF₅O₃
Storage Temperature: Ambient
MDL Number: MFCD11036086
CAS Number: 958451-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR