4-Bromo-3-methoxy-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

153512-1G 153512-500MG 153512-5G 153512-25G
77465-090EA 415.09 USD
77465-090 77477-858 77487-996 77498-688
4-Bromo-3-methoxy-5-(methylthio)benzaldehyde ≥97%
4-Bromo-3-methoxy-5-(methylthio)benzaldehyde

Precaución: For research use only

Formula: C₉H₉BrO₂S
MW: 261.1355 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34843528
CAS Number: 2921905-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR