2-(2-Bromo-6-fluoro-4-methylphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

15316-1G 15316-500MG 15316-5G 15316-25G
77465-086EA 103 USD
77465-086 77477-854 77487-992 77498-684
2-(2-Bromo-6-fluoro-4-methylphenyl)-1,3-dioxolane ≥95%
2-(2-Bromo-6-fluoro-4-methylphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₂
MW: 261.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619362
CAS Number: 2221812-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR