2-(2,6-Dibromophenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

15290-1G 15290-500MG 15290-5G 15290-25G
77465-082EA 151.41 USD
77465-082 77477-850 77487-988 77498-680
2-(2,6-Dibromophenyl)-1,3-dioxolane ≥95%
2-(2,6-Dibromophenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₉H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28976809
CAS Number: 331714-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR