2-Hydroxy-4,6-di(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

15257-1G 15257-500MG 15257-5G 15257-25G
77465-078EA 252.35 USD
77465-078 77477-846 77487-984 77498-676
2-Hydroxy-4,6-di(trifluoromethyl)benzoic acid ≥97%
2-Hydroxy-4,6-di(trifluoromethyl)benzoic acid

Precaución: For research use only

Formula: C₉H₄F₆O₃
MW: 274.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00729117
CAS Number: 180134-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR